siRNA Details
Gene Symbol: Aox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019363.3 | NP_062236.2 | 33 | 3657 | ||
GenBank mRNA | AF110477.1 | - | 33 | 3674 | ||
AF110478.1 | - | 33 | 3658 |

More Information
Other Species Targeted

Gene Ontology Categories:
