siRNA Details
Gene Symbol: Srcin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019378.1 | NP_062251.1 | 2802 | |||
GenBank mRNA | AF156981.1 | - | 2802 | |||
AF156982.1 | - | 2802 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Srcin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019378.1 | NP_062251.1 | 2802 | |||
GenBank mRNA | AF156981.1 | - | 2802 | |||
AF156982.1 | - | 2802 |