siRNA Details
Gene Symbol: Uso1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019379.1 | NP_062252.1 | 9 | 861 | ||
XM_006250750.2 | XP_006250812.1 | 9 | 882 | |||
GenBank mRNA | FQ209746.1 | - | 1 | 50 | ||
U14192.1 | - | 9 | 776 | |||
U15589.1 | - | 9 | 861 |
