siRNA Details
Gene Symbol: Tmbim6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019381.2 | NP_062254.2 | 9, 10 | 780 | ||
XM_006257323.2 | XP_006257385.1 | 9, 10 | 922 | |||
GenBank mRNA | BC058478.1 | - | 9, 10 | 786 | 6888036 | |
FQ217052.1 | - | 9, 10 | 779 | |||
FQ218664.1 | - | 9, 10 | 782 | |||
FQ226858.1 | - | 9, 10 | 809 | |||
FQ229652.1 | - | 9, 10 | 781 | |||
FQ230608.1 | - | 9, 10 | 809 | |||
FQ231817.1 | - | 9, 10 | 787 | |||
X75855.1 | - | 9, 10 | 809 | |||
X75856.1 | - | 8, 9 | 693 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
