siRNA Details
Gene Symbol: Zfp386 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019620.2 | NP_062566.2 | 3 | 1188 | ||
XM_006240685.2 | XP_006240747.1 | 3 | 1345 | |||
GenBank mRNA | BC078841.1 | - | 3 | 1187 | 7115131 | |
U67082.1 | - | 3 | 1177 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
