siRNA Details
Gene Symbol: Pth1r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020073.2 | NP_064458.1 | 12, 13 | 1406 | ||
XM_006243998.2 | XP_006244060.1 | 15, 16 | 1601 | |||
XM_006243999.2 | XP_006244061.1 | 13, 14 | 1593 | |||
XM_006244000.2 | XP_006244062.1 | 14, 15 | 1481 | |||
XM_006244002.2 | XP_006244064.1 | 13, 14 | 1493 | |||
XM_008766651.1 | XP_008764873.1 | 12, 13 | 3505 | |||
GenBank mRNA | L19475.1 | - | 12, 13 | 1406 | ||
M77184.1 | - | 13, 14 | 1433 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Gene Ontology Categories:
