siRNA Details
Gene Symbol: Eif5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020075.1 | NP_064460.1 | 5 | 695 | ||
XM_006240608.1 | XP_006240670.1 | 7 | 958 | |||
XM_006240609.2 | XP_006240671.1 | 6 | 1238 | |||
GenBank mRNA | BC062398.1 | - | 6 | 883 | 5623373 | |
FQ222051.1 | - | 3 | 775 | |||
L11651.1 | - | 5 | 695 |
Gene Symbol: RGD1564400 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006234307.2 | XP_006234369.1 | 1 | 84 |
