siRNA Details
Gene Symbol: Ptprk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029902.2 | NP_001025073.2 | 2 | 227 | ||
XM_008758616.1 | XP_008756838.1 | 2 | 424 | |||
XM_008758617.1 | XP_008756839.1 | 2 | 423 | |||
XM_008758618.1 | XP_008756840.1 | 2 | 423 | |||
XM_008758619.1 | XP_008756841.1 | 2 | 431 | |||
XM_008758620.1 | XP_008756842.1 | 2 | 431 | |||
GenBank mRNA | AB288087.1 | - | 2 | 226 | ||
AB297790.1 | - | 2 | 227 | |||
AF142480.1 | - | 2 | 152 |

More Information
Other Species Targeted

Gene Ontology Categories:
