siRNA Details
Gene Symbol: Parva | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020656.2 | NP_065707.2 | 1 | 127 | ||
GenBank mRNA | AF264765.2 | - | 1 | 127 | ||
FQ228521.1 | - | 187 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
