siRNA Details
Gene Symbol: Cnksr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113366.1 | NP_001106837.1 | 1, 2 | 245 | ||
NM_021686.3 | NP_067718.1 | 1, 2 | 245 | |||
XM_008773224.1 | XP_008771446.1 | 1, 2 | 404 | |||
GenBank mRNA | AF102853.1 | - | 1, 2 | 72 | ||
AF102854.1 | - | 1, 2 | 534 |
