siRNA Details
Gene Symbol: Arhgef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF314539.1 | - | 107 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001130152, NM_001130153, NM_008488, NM_001130150, NM_001130151, and others.
