siRNA Details
Gene Symbol: Nucb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021663.2 | NP_067695.1 | 5 | 520 | ||
XM_006230095.2 | XP_006230157.1 | 4 | 428 | |||
GenBank mRNA | AF238223.1 | - | 3 | 307 | ||
AF250142.1 | - | 4 | 407 | |||
BC061778.1 | - | 5 | 520 | 5621784 |
