siRNA Details
Gene Symbol: Prom1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110137.1 | NP_001103607.1 | 2, 3 | 432 | ||
NM_021751.2 | NP_068519.2 | 3, 4 | 499 | |||
XM_008770215.1 | XP_008768437.1 | 3, 4 | 835 | |||
XM_008770218.1 | XP_008768440.1 | 3, 4 | 540 | |||
GenBank mRNA | AF263368.1 | - | 2, 3 | 466 | ||
AF386758.1 | - | 2, 3 | 378 | |||
AY262731.1 | - | 2, 3 | 378 |
