siRNA Details
Gene Symbol: Birc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021752.2 | NP_068520.2 | 1 | 1121 | ||
XM_006242494.2 | XP_006242556.1 | 2 | 1598 | |||
XM_006242495.2 | XP_006242557.1 | 2 | 1799 | |||
GenBank mRNA | AF183431.1 | - | 1 | 479 | ||
AF190020.1 | - | 1 | 1493 | |||
BC062055.1 | - | 1 | 1121 | 5598786 |
