siRNA Details
Gene Symbol: Epn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033914.1 | NP_001029086.1 | 2 | 588 | ||
NM_021852.2 | NP_068624.2 | 2 | 588 | |||
XM_006246521.2 | XP_006246583.1 | 1 | 336 | |||
XM_006246522.2 | XP_006246584.1 | 3 | 670 | |||
XM_006246523.2 | XP_006246585.1 | 1 | 336 | |||
GenBank mRNA | AF096269.1 | - | 1 | 329 | ||
BC094524.1 | - | 2 | 567 | 7318094 | ||
FQ211574.1 | - | 3 | 670 | |||
FQ211939.1 | - | 2 | 612 |

More Information
Other Species Targeted

Gene Ontology Categories:
