siRNA Details
Gene Symbol: Exoc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022204.3 | NP_071540.1 | 14 | 1700 | ||
GenBank mRNA | BC128739.1 | - | 14 | 1703 | 8362342 | |
FQ217124.1 | - | 13 | 1534 | |||
U79417.1 | - | 14 | 1472 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_207214, AK170148, AK164067, AK150297, AK167145, and others.
Gene Ontology Categories:
