siRNA Details
Gene Symbol: Ppp2r2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022209.3 | NP_071545.2 | 5 | 797 | ||
GenBank mRNA | BC078834.1 | - | 8 | 1006 | 7114294 | |
D38260.1 | - | 5 | 592 | |||
FQ212581.1 | - | 3 | 317 |

More Information
Other Species Targeted

Gene Ontology Categories:
