siRNA Details
Gene Symbol: Insrr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006232749.2 | XP_006232811.2 | 4 | 1557 | ||
XM_008761207.1 | XP_008759429.1 | 4 | 1557 | |||
GenBank mRNA | D13966.1 | - | 4 | 1089 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
