siRNA Details
Gene Symbol: Kcne3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022235.2 | NP_071571.1 | 2 | 229 | ||
XM_006229785.2 | XP_006229847.1 | 3 | 2221 | |||
XM_006229786.2 | XP_006229848.1 | 3 | 1076 | |||
GenBank mRNA | AJ271742.1 | - | 3 | 364 | ||
BC086406.1 | - | 2 | 229 | 7306835 |
