siRNA Details
Gene Symbol: Fbxl20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022272.1 | NP_071608.1 | 2, 3 | 112 | ||
XM_006247421.2 | XP_006247483.1 | 2, 3 | 414 | |||
XM_008768110.1 | XP_008766332.1 | 2, 3 | 416 | |||
XM_008768111.1 | XP_008766333.1 | 2, 3 | 451 | |||
GenBank mRNA | AF182443.1 | - | 2, 3 | 112 |
