siRNA Details
Gene Symbol: Pde10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022236.1 | NP_071572.1 | 2 | 378 | ||
GenBank mRNA | AB027155.1 | - | 2 | 378 | ||
AB027156.1 | - | 713 | ||||
AY462091.1 | - | 344 | ||||
AY462092.1 | - | 452 | ||||
AY462093.1 | - | 490 | ||||
AY462095.1 | - | 625 |
