siRNA Details
Gene Symbol: Casp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022260.3 | NP_071596.1 | 5, 6 | 691 | ||
XM_008760546.1 | XP_008758768.1 | 6, 7 | 1284 | |||
XM_008760547.1 | XP_008758769.1 | 5, 6 | 1666 | |||
XM_008760548.1 | XP_008758770.1 | 5, 6 | 993 | |||
XM_008760549.1 | XP_008758771.1 | 6, 7 | 873 | |||
XM_008760550.1 | XP_008758772.1 | 5, 6 | 801 | |||
GenBank mRNA | AF072124.1 | - | 5, 6 | 692 | ||
BC070936.1 | - | 5, 6 | 692 | 7099111 |
