siRNA Details
Gene Symbol: Dlg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022282.1 | NP_071618.1 | 5 | 603 | ||
XM_008759648.1 | XP_008757870.1 | 5 | 750 | |||
GenBank mRNA | U49049.1 | - | 5 | 603 | ||
U50717.1 | - | 5 | 327 |

More Information
Other Species Targeted

Gene Ontology Categories:
