siRNA Details
Gene Symbol: Tuba1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022298.1 | NP_071634.1 | 3 | 362 | ||
GenBank mRNA | BC062238.1 | - | 3 | 396 | 6922126 | |
BC078830.1 | - | 3 | 375 | 7115494 | ||
FQ210110.1 | - | 3 | 394 | |||
FQ212756.1 | - | 3 | 328 | |||
FQ213186.1 | - | 3 | 394 | |||
V01227.1 | - | 3 | 362 |
Gene Symbol: LOC100909441 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749354.3 | XP_003749402.1 | 3 | 389 |
Gene Symbol: Tuba1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044270.2 | NP_001037735.1 | 3 | 395 | ||
GenBank mRNA | BC060572.1 | - | 3 | 399 | 6920989 | |
BC076379.1 | - | 3 | 381 | 7106502 | ||
FQ212426.1 | - | 3 | 390 | |||
FQ212740.1 | - | 3 | 390 | |||
FQ213098.1 | - | 3 | 395 | |||
FQ213226.1 | - | 3 | 390 | |||
FQ213439.1 | - | 3 | 390 | |||
FQ213947.1 | - | 3 | 390 | |||
FQ214075.1 | - | 3 | 393 | |||
FQ214100.1 | - | 3 | 390 | |||
FQ214301.1 | - | 3 | 393 | |||
FQ223081.1 | - | 3 | 393 | |||
FQ225046.1 | - | 3 | 395 | |||
FQ225339.1 | - | 3 | 390 | |||
FQ225425.1 | - | 3 | 390 | |||
FQ225650.1 | - | 3 | 390 | |||
FQ228903.1 | - | 177 | ||||
FQ231550.1 | - | 3 | 390 | |||
FQ232160.1 | - | 2 | 300 | |||
FQ232527.1 | - | 3 | 394 | |||
FQ233000.1 | - | 3 | 392 | |||
FQ233566.1 | - | 3 | 393 | |||
FQ234135.1 | - | 3 | 390 |
Gene Symbol: Tuba1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011995.1 | NP_001011995.1 | 3 | 378 | ||
GenBank mRNA | BC078829.1 | - | 3 | 378 | 7112420 | |
FQ221879.1 | - | 2 | 168 | |||
FQ225183.1 | - | 3 | 399 | |||
FQ225366.1 | - | 3 | 398 | |||
FQ226218.1 | - | 3 | 399 | |||
FQ235315.1 | - | 3 | 399 |

More Information
Other Species Targeted

Gene Ontology Categories:
