siRNA Details
Gene Symbol: Card9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022303.1 | NP_071639.1 | 3 | 395 | ||
XM_008761625.1 | XP_008759847.1 | 3 | 1583 | |||
XM_008761626.1 | XP_008759848.1 | 3 | 1583 | |||
GenBank mRNA | AF311288.1 | - | 3 | 395 |
