siRNA Details
Gene Symbol: Pafah1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022387.3 | NP_071782.1 | 4, 5 | 421 | ||
XM_006242957.2 | XP_006243019.1 | 4, 5 | 887 | |||
XM_006242958.2 | XP_006243020.1 | 4, 5 | 420 | |||
XM_008766172.1 | XP_008764394.1 | 4, 5 | 651 | |||
GenBank mRNA | AF016048.1 | - | 4, 5 | 384 | ||
BC081714.1 | - | 4, 5 | 421 | 7108923 | ||
FQ212439.1 | - | 4, 5 | 708 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008775, AK153424, AK012164, BC002037, AK012116, and others.
Gene Ontology Categories:
