siRNA Details
Gene Symbol: Casp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022260.3 | NP_071596.1 | 6 | 771 | ||
XM_008760546.1 | XP_008758768.1 | 7 | 1364 | |||
XM_008760547.1 | XP_008758769.1 | 6 | 1746 | |||
XM_008760548.1 | XP_008758770.1 | 6 | 1073 | |||
XM_008760549.1 | XP_008758771.1 | 7 | 953 | |||
XM_008760550.1 | XP_008758772.1 | 6 | 881 | |||
GenBank mRNA | AF072124.1 | - | 6 | 772 | ||
BC070936.1 | - | 6 | 772 | 7099111 |
