siRNA Details
Gene Symbol: Dlg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022282.1 | NP_071618.1 | 23 | 2748 | ||
XM_008759648.1 | XP_008757870.1 | 24 | 2991 | |||
XM_008759649.1 | XP_008757871.1 | 12 | 3429 | |||
GenBank mRNA | U49049.1 | - | 23 | 2748 | ||
U50717.1 | - | 22 | 2406 |

More Information
Other Species Targeted

Gene Ontology Categories:
