siRNA Details
Gene Symbol: Prl8a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022537.1 | NP_071982.1 | 5, 6 | 603 | ||
XM_006253909.1 | XP_006253971.1 | 5, 6 | 577 | |||
GenBank mRNA | AB000107.1 | - | 5, 6 | 603 | ||
BC098917.1 | - | 5, 6 | 583 | 7445772 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
