siRNA Details
Gene Symbol: Entpd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022587.1 | NP_072109.1 | 7 | 936 | ||
GenBank mRNA | U81295.1 | - | 7 | 936 |
Gene Symbol: LOC103689954 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760464.1 | XP_008758686.1 | 872 |
