siRNA Details
Gene Symbol: Golga2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022596.2 | NP_072118.2 | 26 | 3029 | ||
XM_006233992.2 | XP_006234054.1 | 27 | 3118 | |||
XM_006233993.2 | XP_006234055.1 | 27 | 3085 | |||
XM_006233994.2 | XP_006234056.1 | 27 | 3082 | |||
XM_006233995.2 | XP_006234057.1 | 26 | 3037 | |||
XM_006233996.2 | XP_006234058.1 | 26 | 3038 | |||
XM_006233997.2 | XP_006234059.1 | 26 | 3004 | |||
XM_006233998.2 | XP_006234060.1 | 26 | 3001 | |||
XM_008761774.1 | XP_008759996.1 | 25 | 2942 | |||
GenBank mRNA | U35022.2 | - | 26 | 3032 |
