siRNA Details
Gene Symbol: Slit2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022632.2 | NP_072154.2 | 19 | 2557 | ||
XM_008770210.1 | XP_008768432.1 | 16 | 1692 |

More Information
Other Species Targeted

Gene Ontology Categories:
