siRNA Details
Gene Symbol: EPHA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017000534.1 | XP_016856023.1 | 3 | 626 | ||
XM_017000535.1 | XP_016856024.1 | 3 | 586 | |||
XM_017000536.1 | XP_016856025.1 | 2 | 434 | |||
XM_017000537.1 | XP_016856026.1 | 3 | 528 | |||
GenBank mRNA | BC037166.2 | - | 3 | 474 | 5090654 | |
CB991812.1 | - | 3 | 538 | |||
EU826606.1 | - | 3 | 356 | |||
M59371.1 | - | 3 | 469 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
