siRNA Details
Gene Symbol: B4galnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022860.2 | NP_074051.1 | 3 | 484 | ||
XM_006241497.2 | XP_006241559.1 | 3 | 1011 | |||
XM_008765419.1 | XP_008763641.1 | 3 | 2840 | |||
XM_008765420.1 | XP_008763642.1 | 2 | 1161 | |||
GenBank mRNA | BC081799.1 | - | 3 | 484 | 7133837 | |
D17809.1 | - | 3 | 373 |
