siRNA Details
Gene Symbol: Tpi1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022922.2 | NP_075211.2 | 4 | 436 | ||
GenBank mRNA | BC061781.1 | - | 4 | 411 | ||
BC126087.1 | - | 4 | 436 | 8368624 | ||
FQ214607.1 | - | 4 | 521 | |||
FQ214670.1 | - | 4 | 444 | |||
FQ214880.1 | - | 4 | 449 | |||
FQ215213.1 | - | 3 | 351 | |||
FQ215529.1 | - | 4 | 448 | |||
FQ215606.1 | - | 3 | 324 | |||
FQ215810.1 | - | 4 | 449 | |||
FQ216226.1 | - | 3 | 374 | |||
FQ216307.1 | - | 4 | 335 | |||
FQ216584.1 | - | 4 | 448 | |||
FQ223237.1 | - | 4 | 450 | |||
FQ224880.1 | - | 4 | 447 | |||
FQ224920.1 | - | 4 | 451 | |||
FQ224970.1 | - | 4 | 444 | |||
L36250.1 | - | 4 | 436 |
Gene Symbol: LOC100911515 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750654.3 | XP_003750702.2 | 4 | 592 |
Gene Symbol: LOC500959 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033072.1 | NP_001028244.1 | 1 | 433 | ||
GenBank mRNA | AY461585.1 | - | 1 | 433 |

More Information
Other Species Targeted

Gene Ontology Categories:
