siRNA Details
Gene Symbol: Galnt7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022926.1 | NP_075215.1 | 1, 2 | 184 | ||
XM_006253083.1 | XP_006253145.1 | 1, 2 | 209 | |||
GenBank mRNA | AF076167.1 | - | 1, 2 | 184 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_144731, NM_001167981, BC052461, BY167413, AK041791, and others.
Gene Ontology Categories:
