siRNA Details
Gene Symbol: Snap23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022689.2 | NP_073180.1 | 5 | 328 | ||
GenBank mRNA | AF052596.1 | - | 180 | |||
BC127494.1 | - | 5 | 328 | 8361815 |

More Information
Other Species Targeted

Gene Ontology Categories:
