siRNA Details
Gene Symbol: Mid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022927.1 | NP_075216.1 | 2 | 786 | ||
XM_006256825.2 | XP_006256887.1 | 2 | 2070 | |||
GenBank mRNA | AF186461.1 | - | 2 | 786 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_183151, NM_010797, Y14848, AY540038, AF026565, and others.
Gene Ontology Categories:
