siRNA Details
Gene Symbol: Mug1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023103.1 | NP_075591.1 | 2 | 302 | ||
XM_006237282.2 | XP_006237344.1 | 2 | 342 | |||
GenBank mRNA | M22358.1 | - | 2 | 330 | ||
M22360.1 | - | 1 | 16 | |||
X52984.1 | - | 2 | 302 |
Gene Symbol: Cpamd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271376.1 | NP_001258305.1 | 251 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
