siRNA Details
Gene Symbol: Psme4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025140.2 | NP_001020311.2 | 3 | 654 | ||
XM_008770467.1 | XP_008768689.1 | 3 | 904 |

More Information
Other Species Targeted

Gene Ontology Categories:
