siRNA Details
Gene Symbol: Fgfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024146.1 | NP_077060.1 | 3 | 386 | ||
XM_006253323.2 | XP_006253385.1 | 3 | 394 | |||
XM_006253324.2 | XP_006253386.1 | 4 | 607 | |||
XM_006253325.2 | XP_006253387.1 | 2 | 127 | |||
XM_006253326.2 | XP_006253388.1 | 2 | 127 | |||
XM_006253327.1 | XP_006253389.1 | 3 | 414 | |||
GenBank mRNA | D12498.1 | - | 3 | 386 | ||
S54008.1 | - | 3 | 419 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
