siRNA Details
Gene Symbol: EPHA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288629.1 | NP_001275558.1 | 3, 4 | 1066 | ||
NM_004440.3 | NP_004431.1 | 3, 4 | 1066 | |||
XM_005248669.2 | XP_005248726.1 | 3, 4 | 1123 | |||
XM_005248671.2 | XP_005248728.1 | 3, 4 | 1123 | |||
XM_017010365.1 | XP_016865854.1 | 3, 4 | 1123 | |||
XM_017010366.1 | XP_016865855.1 | 3, 4 | 1123 | |||
GenBank mRNA | AB209269.1 | - | 3, 4 | 1053 | ||
AK313529.1 | - | 3, 4 | 1038 | |||
BC126125.1 | - | 3, 4 | 940 | 8991841 | ||
BC126151.1 | - | 3, 4 | 940 | 8991867 | ||
BC143857.1 | - | 3, 4 | 940 | 9052373 | ||
BC143858.1 | - | 3, 4 | 940 | 9052374 | ||
L36642.1 | - | 3, 4 | 1010 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
