siRNA Details
Gene Symbol: Mug1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023103.1 | NP_075591.1 | 3 | 359 | ||
XM_006237282.2 | XP_006237344.1 | 3 | 399 | |||
GenBank mRNA | M22358.1 | - | 3 | 387 | ||
M22360.1 | - | 2 | 73 | |||
X52984.1 | - | 3 | 359 |
Gene Symbol: A1i3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037975.2 | NP_001033064.2 | 3 | 387 | ||
XM_006237281.2 | XP_006237343.1 | 3 | 399 | |||
GenBank mRNA | J03552.1 | - | 3 | 387 |
Gene Symbol: Cpamd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271376.1 | NP_001258305.1 | 308 |
Gene Symbol: LOC100911784 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763278.1 | XP_008761500.1 | 1 | 71 |
Gene Symbol: Mug2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002826.1 | NP_001002826.1 | 305 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
