siRNA Details
Gene Symbol: Faah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024132.3 | NP_077046.1 | 2, 3 | 378 | ||
GenBank mRNA | BC091148.1 | - | 379 | 7370226 | ||
CO569058.1 | - | 378 | ||||
U72497.1 | - | 366 |
Gene Symbol: LOC100911581 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750001.3 | XP_003750049.1 | 2, 3 | 723 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_010173, AK138481, U82536, AK080603, AK004985, and others.
Gene Ontology Categories:
