siRNA Details
Gene Symbol: ELF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114309.1 | NP_001107781.1 | 9 | 2315 | ||
NM_004433.4 | NP_004424.3 | 9 | 2201 | |||
GenBank mRNA | AF017307.1 | - | 9 | 2245 | ||
CV570925.1 | - | 1 | 18 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
