siRNA Details
Gene Symbol: Golph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023977.2 | NP_076467.2 | 2, 3 | 474 | ||
GenBank mRNA | AF311954.1 | - | 2, 3 | 365 | ||
BC092568.1 | - | 2, 3 | 474 | 7307277 |

More Information
Other Species Targeted

Gene Ontology Categories:
