siRNA Details
Gene Symbol: Apaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023979.1 | NP_076469.1 | 7 | 1495 | ||
XM_008765263.1 | XP_008763485.1 | 4 | 1287 | |||
GenBank mRNA | AF218388.1 | - | 7 | 950 | ||
AF320222.1 | - | 7 | 1495 |
