siRNA Details
Gene Symbol: Psme4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025140.2 | NP_001020311.2 | 21 | 2805 | ||
XM_008770467.1 | XP_008768689.1 | 22 | 3121 | |||
GenBank mRNA | DY314798.1 | - | 3 | 169 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
