siRNA Details
Gene Symbol: Notch2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024358.1 | NP_077334.1 | 3 | 683 | ||
GenBank mRNA | M93661.1 | - | 3 | 683 |

More Information
Other Species Targeted

Gene Ontology Categories:
