siRNA Details
Gene Symbol: Hr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024364.3 | NP_077340.3 | 4 | 1069 | ||
XM_006252283.1 | XP_006252345.1 | 1 | 296 | |||
XM_008770833.1 | XP_008769055.1 | 1 | 296 | |||
GenBank mRNA | U71293.1 | - | 1 | 685 |
